Learn More
Medchemexpress LLC HY-115576 5mg Medchemexpress, P62-mediated mitophagy inducer CAS:1809031-84-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1155765MG
Encompass_Preferred
Medchemexpress, HY-115576 5mg P62-mediated mitophagy inducer CAS:1809031-84-2 P62-mediated mitophagy inducer is a mitophagy regulator which activates mitophagy without recruiting Parkin or collapsing ΔΨm and retains activity in cells devoid of a fully functional PINK1/Parkin pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9395