Learn More
Selleck Chemical LLC P62-mediated mitophagy inducer
SDP
Catalog No. 503135778
Encompass_Preferred
P62-mediated mitophagy inducer (PMI) is a P62-mediated activator of mitophagy that functions independently of parkin recruitment or mitochondrial membrane potential ( m) collapse It remains effective even in cells lacking a fully functional PINK1/Parkin pathway PMI stabilizes Nrf2 and enhances p62 expression thereby activating mitophagy
Catalog No. 50-313-5778 Supplier Selleck Chemical LLC Supplier No. E14621G
P62-mediated mitophagy inducer (PMI) is a P62-mediated activator of mitophagy that functions independently of parkin recruitment or mitochondrial membrane potential ( m) collapse It remains effective even in cells lacking a fully functional PINK1/Parkin pathway PMI stabilizes Nrf2 and enhances p62 expression thereby activating mitophagy