Learn More
Selleck Chemical LLC P62-mediated mitophagy inducer
Supplier: Selleck Chemical LLC E146225MG
P62-mediated mitophagy inducer (PMI) is a P62-mediated activator of mitophagy that functions independently of parkin recruitment or mitochondrial membrane potential ( m) collapse It remains effective even in cells lacking a fully functional PINK1/Parkin pathway PMI stabilizes Nrf2 and enhances p62 expression thereby activating mitophagy