Learn More
Enzo Life Sciences p62-derived UBA domain, (agarose immobilized) (0.5ML)
SDP
Catalog No. 502012294
Encompass_Preferred
Formulation: The resin is supplied as a 50% slurry in 25 mM TRIS-HCl, pH 7.5, containing 0.01% sodium azide. Source: Synthesized. Long Term Storage: +4°C.
Catalog No. 50-201-2294 Supplier Enzo Life Sciences Supplier No. BMLUW90100500
Formulation: The resin is supplied as a 50% slurry in 25 mM TRIS-HCl, pH 7.5, containing 0.01% sodium azide. Source: Synthesized. Long Term Storage: +4°C.