Learn More
Cayman Chemical p62SQSTM1 PolYCNl ntIbod 500uL
SDP
Catalog No. 502935615
Encompass_Preferred
A B2-AR agonist selectively binds to B1- over B2-ARs (Kds 2.9 and 113 nM respectively) as well as B3-adrenergic B2 bradykinin NK1 and NK2 receptors up to 3 M induces cAMP accumulation in cultured human bronchial epithelial cells relaxes guinea pig tracheal strips precontracted with carbamoylcholine ovalbumin or histamine (pD2s 8.4 9.5 and 9.5 respectively) reverses histamine- and ovalbumin-induced bronchoconstriction in guinea pigs (ED50s 1 and 40 nmol/kg respectively)
Catalog No. 50-293-5615 Supplier Cayman Chemical Supplier No. 24693500 UL
A B2-AR agonist selectively binds to B1- over B2-ARs (Kds 2.9 and 113 nM respectively) as well as B3-adrenergic B2 bradykinin NK1 and NK2 receptors up to 3 M induces cAMP accumulation in cultured human bronchial epithelial cells relaxes guinea pig tracheal strips precontracted with carbamoylcholine ovalbumin or histamine (pD2s 8.4 9.5 and 9.5 respectively) reverses histamine- and ovalbumin-induced bronchoconstriction in guinea pigs (ED50s 1 and 40 nmol/kg respectively)