Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 8626MSM6P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: TP63/1786. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HEK293 cells or Prostate Carcinoma or Lung or bladder squamous cell carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human p63 protein fragment (aa3-106). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 63kDa.Chromosome: 3q28. Gene Symbol: TP63. Gene ID: 8626. Ensemble: ENSG00000073282. Uniprot: NP_001316893.
Catalog No. 50-188-5310