Learn More
Perkin Elmer US LLC P7+ prepFAST sample valve
Catalog No. 502112202
Encompass_Preferred
P7+ prepFAST sample valve with CTFE stator and 1mm PFA rotor. Includes center and side mixing ports.
Catalog No. 50-211-2202 Supplier Perkin Elmer US LLC Supplier No. N0777750
P7+ prepFAST sample valve with CTFE stator and 1mm PFA rotor. Includes center and side mixing ports.