Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P70 S6 Kinase beta/SRK (phospho S371) Antibody (YA1890) | 50 UL
SDP

Catalog No. 5000441315
Encompass_Preferred

P70 S6 Kinase beta/SRK (phospho S371) Antibody (YA1890) is a Rabbit-derived, non-conjugated IgG monoclonal antibody that specifically targets P70 S6 Kinase beta/SRK (phospho S371). It is supplied as a liquid formulation for research use.

  • Rabbit-derived
  • Non-conjugated
  • IgG monoclonal antibody
  • Targets P70 S6 Kinase beta/SRK (phospho S371)
  • Application: Western blotting (WB)
  • Reactivity: Human, mouse, rat
  • Formulation: 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Storage: -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipping: With blue ice

Catalog No. 50-004-41315 Supplier Medchemexpress LLC Supplier No. HYP8214550UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P70 S6 Kinase beta/SRK (phospho S371) Antibody (YA1890) is a Rabbit-derived, non-conjugated IgG monoclonal antibody that specifically targets P70 S6 Kinase beta/SRK (phospho S371). It is supplied as a liquid formulation for research use.

  • Rabbit-derived
  • Non-conjugated
  • IgG monoclonal antibody
  • Targets P70 S6 Kinase beta/SRK (phospho S371)
  • Application: Western blotting (WB)
  • Reactivity: Human, mouse, rat
  • Formulation: 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Storage: -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipping: With blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.