Learn More
Enzo Life Sciences p70 S6 kinase (human), (recombinant) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE3450005
Encompass_Preferred
Alternative name: p70 S6K. Formulation: Liquid. In 50mM sodium phosphate, pH 7.0, containing 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, and 25% glycerol. Source: Produced in insect cells. Active, full length p70 S6K. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-1989