Learn More
Instech Kapton strip kit for Instech P720 peristaltic pumps. 10 strips, 3 pcs gripping paper, clip, tool. Can improve accuracy and prevent wear of pump tubes.
SDP
Catalog No. 5081172
Encompass
The Instech P720/KSK is a Kapton strip kit designed for the maintenance and repair of the P720 series of peristaltic pumps. These strips are applied to the pump head to provide a smooth, low-friction surface for the tubing and rollers, ensuring consistent flow rates and extending the life of the pump components.
Features
  • Type: Pump maintenance and repair kit
  • Contents: Includes 10 Kapton strips and 3 pieces of gripping paper
  • Tools Included: One clip and one specialized application tool
  • Compatibility: Designed for use with P720/10K, P720, and P720/66 pump heads
  • Function: Replaces worn or damaged friction surfaces to maintain pump accuracy
  • Application: Used when the original Kapton surface becomes wrinkled or starts to peel
Catalog No. 50-811-72 Supplier Instech Supplier No. P720/KSK
 Restricted Item
Add to Cart
Add to Cart
The Instech P720/KSK is a Kapton strip kit designed for the maintenance and repair of the P720 series of peristaltic pumps. These strips are applied to the pump head to provide a smooth, low-friction surface for the tubing and rollers, ensuring consistent flow rates and extending the life of the pump components.
Features
  • Type: Pump maintenance and repair kit
  • Contents: Includes 10 Kapton strips and 3 pieces of gripping paper
  • Tools Included: One clip and one specialized application tool
  • Compatibility: Designed for use with P720/10K, P720, and P720/66 pump heads
  • Function: Replaces worn or damaged friction surfaces to maintain pump accuracy
  • Application: Used when the original Kapton surface becomes wrinkled or starts to peel