Learn More
Instech Fittings to allow straight-through tube sets to be used with Instech P720 peristaltic pumps. Set of 2. Use with P720/TS-020S, -031S, -062S or -093S tube sets.
SDP
Catalog No. 5081173
Encompass
The Instech P720/NCA is a pair of nylon connectors designed to secure straight-through tube sets into the pump head of any P720 series peristaltic pump. These fittings are necessary when using custom or non-barbed tubing configurations where standard pre-assembled connectors are not used.
Features
  • Type: Tube set fittings for straight-through configurations
  • Construction: Durable nylon
  • Quantity: Supplied as a pair (2 pieces)
  • Compatibility: Required for P720 straight-through tube sets in sizes 0.020in (0.5mm), 0.031in (0.8mm), 0.062in (1.6mm), and 0.093in (2.4mm) ID
  • Function: Securely holds the tubing in place within the pump head to ensure accurate flow
  • Application: Ideal for setups where a continuous fluid path is needed without the potential dead volume of intermediate barbed connectors
Catalog No. 50-811-73 Supplier Instech Supplier No. P720/NCA
 Restricted Item
Add to Cart
Add to Cart
The Instech P720/NCA is a pair of nylon connectors designed to secure straight-through tube sets into the pump head of any P720 series peristaltic pump. These fittings are necessary when using custom or non-barbed tubing configurations where standard pre-assembled connectors are not used.
Features
  • Type: Tube set fittings for straight-through configurations
  • Construction: Durable nylon
  • Quantity: Supplied as a pair (2 pieces)
  • Compatibility: Required for P720 straight-through tube sets in sizes 0.020in (0.5mm), 0.031in (0.8mm), 0.062in (1.6mm), and 0.093in (2.4mm) ID
  • Function: Securely holds the tubing in place within the pump head to ensure accurate flow
  • Application: Ideal for setups where a continuous fluid path is needed without the potential dead volume of intermediate barbed connectors