Learn More
Medchemexpress LLC P7C3-OMe | 313268-18-7 | 98.3% | 10 MG
SDP
Catalog No. 5000338737
Encompass_Preferred
P7C3-OMe is a pro-neurogenic compound with a molecular formula of C22H20Br2N2O2 and a molecular weight of 504.21. It has therapeutic benefits in neuropsychiatric and neurodegenerative diseases.
  • Pro-neurogenic compound
  • Potential therapeutic benefits in neuropsychiatric diseases
  • Potential therapeutic benefits in neurodegenerative diseases
  • R-enantiomer exhibits enhanced activity
Catalog No. 50-003-38737 Supplier Medchemexpress LLC Supplier No. HY12885610MG
P7C3-OMe is a pro-neurogenic compound with a molecular formula of C22H20Br2N2O2 and a molecular weight of 504.21. It has therapeutic benefits in neuropsychiatric and neurodegenerative diseases.
  • Pro-neurogenic compound
  • Potential therapeutic benefits in neuropsychiatric diseases
  • Potential therapeutic benefits in neurodegenerative diseases
  • R-enantiomer exhibits enhanced activity