Learn More
Abcam P7C3, pro-neurogenic and neuroprotective compound, 50MG
Catalog No. 502706817
Encompass_Preferred
MW 474.2 Da, Purity >99%. Pro-neurogenic and neuroprotective compound. Blocks apoptosis of newborn neurons of the dentate gyrus. Also protects mature neurons. Enhances neurogenesis and additionally improves cognitive behaviour. Blood-brain barrier permeable. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6817 Supplier Abcam Supplier No. AB14118850MG
MW 474.2 Da, Purity >99%. Pro-neurogenic and neuroprotective compound. Blocks apoptosis of newborn neurons of the dentate gyrus. Also protects mature neurons. Enhances neurogenesis and additionally improves cognitive behaviour. Blood-brain barrier permeable. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement