Learn More
BD Cell Analysis 3P BD P800 Blood Collection System, Stabilizes Plasma Markers, 2 ml Draw Volume
Supplier:  BD Cell Analysis 3P 366420
Encompass
The BD P800 Blood Collection System enhances accuracy in measuring plasma metabolic markers. Designed for research, it stabilizes critical biomarkers at the point of collection using a proprietary combination of protease inhibitors and K2EDTA. This ensures sample integrity for immediate analysis and transport, making it ideal for preserving sensitive markers like GLP-1 and ghrelin. Its innovative design, along with features like a BD Hemogard™ Closure, sets it apart from standard blood collection tubes.
Catalog No. NC3588099