Learn More
LAB ALLEY Lab Alley P81 Ion Exchange Cellulose Chromatography Paper
SDP
Catalog No. NC3306482
Encompass
P81 Ion Exchange Cellulose Chromatography Paper is a high-performance cation-exchange medium designed for efficient separation and analysis of biomolecules including proteins peptides and nucleotides. Manufactured from high-quality cellulose fibers this chromatography paper is chemically treated to introduce strong cation-exchange properties making it ideal for electrophoretic and chromatographic applications.
Catalog No. NC3306482 Supplier LAB ALLEY Supplier No. 057172B
P81 Ion Exchange Cellulose Chromatography Paper is a high-performance cation-exchange medium designed for efficient separation and analysis of biomolecules including proteins peptides and nucleotides. Manufactured from high-quality cellulose fibers this chromatography paper is chemically treated to introduce strong cation-exchange properties making it ideal for electrophoretic and chromatographic applications.