Learn More
Cell Signaling Technology p90RSK Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502730010
Encompass
The p90RSK Antibody Sampler Kit provides an economical means of evaluating levels of p90RSK protein phosphorylated at specific sites, as well as total p90RSK protein levels. The kit contains enough primary and secondary antibody to perform two Western blot experiments.
  • Includes antibodies for the detection of p90RSK phosphorylated at Thr359, Ser380, and Thr573, plus a total p90RSK antibody
  • Contains HRP-linked secondary antibody for chemiluminescent detection
  • Supports Western blotting applications
  • Validated for reactivity with human, mouse, and rat samples
Catalog No. 50-273-0010 Supplier Cell Signaling Technology Supplier No. 9326T
The p90RSK Antibody Sampler Kit provides an economical means of evaluating levels of p90RSK protein phosphorylated at specific sites, as well as total p90RSK protein levels. The kit contains enough primary and secondary antibody to perform two Western blot experiments.
  • Includes antibodies for the detection of p90RSK phosphorylated at Thr359, Ser380, and Thr573, plus a total p90RSK antibody
  • Contains HRP-linked secondary antibody for chemiluminescent detection
  • Supports Western blotting applications
  • Validated for reactivity with human, mouse, and rat samples