Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ P93 PRO Catheterization Simulator Set
SDPSCI_ED

Catalog No. S37619 Shop All 3B Scientific Products
Change view
Click to view available options
P93 PRO Male and Female catheterization simulator set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S37619 P93 PRO Male and Female catheterization simulator set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37619 Supplier 3B Scientific™ Supplier No. P93PS

missing translation for 'validMainframeMsg'

Includes

Realistic lower abdominal model, Catheterization set with transparent bladder, Bottle and tube connectors and Carry case

Procedures can be realistically demonstrated, practiced and assessed. 3B Scientific™ P93 PRO Catheterization Simulator Set has suprapubic catheter which students can practice along with the trainer.

  • Female and male catheterization with realistic resistance
  • Three levels of adjustable narrowing of urethra
  • Soft and movable foreskin and labia
  • Anatomically realistic pelvic structure
  • Liquid outflows if catheterization is successfully carried out
  • Transparent bladder can be checked, and the abdominal wall can be removed
  • Includes suprapubic catheter for practicing, cleaning and caring
  • Stands securely on table with the help of non-slip feet
  • Magnetic connectors for quick set-up and dismantling
  • Easy to clean, maintain and can be completely taken apart
  • Sponge present inside prevents build-up of moisture

Warranty and Services

3 Year

Specifications

Product Type PRO Male and Female Simulator Set
For Use With (Application) Catheterization
Dimensions (L x W x H) 50 x 42 x 30 cm
Weight (English) 7.71 lb.
Weight (Metric) 3.5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.