Learn More
Cellvis 96 Well glass bottom plate with high performance #1.5 cover glass, 20/cs
Catalog No. NC0536760
Encompass
96 well glass bottom plate. Black polystyrene frame with high performance #1.5 cover glass (0.170+/-0.005mm), with lid, Individually packed.
Catalog No. NC0536760 Supplier Cellvis Supplier No. P961.5HN
96 well glass bottom plate. Black polystyrene frame with high performance #1.5 cover glass (0.170+/-0.005mm), with lid, Individually packed.