Learn More
Carbosynth LLC. Mouse monoclonal p97 ATPase antibody
SDP
Supplier:  Carbosynth LLC. 10RP104A500UL
Encompass_Preferred
p97 ATPase / VCP (also described as valosin-containing protein) is implicated in homotypic membrane fusion events and is related to the N-ethyl maleimide-sensitive fusion protein NSF/Sec18p. The antibody reacts with the p97 subunit of 15S Mg2+-ATPase. p97/VCP is involved in ubiquitin-proteasome dependent protein degradation processes and involvement was also found e.g. in the formation of neuronal inclusion bodies in neuro-degenerative diseseases (e.g. Parkinson¿s disease).
Catalog No. 50-125-2772