Learn More
Miles Scientific PA/20X20 GF 1000U 25/BX
SDP
Catalog No. NC9271030
Encompass_Preferred
Silica Gel GF 1000micrometer 20x20cm w/Preadsorbent Zone 25 plates/box
Catalog No. NC9271030 Supplier Miles Scientific Supplier No. P32013
Silica Gel GF 1000micrometer 20x20cm w/Preadsorbent Zone 25 plates/box