Learn More
Enzo Life Sciences Phosphatidic acid, dioleoyl (10mg). CAS: 108392-02-5
SDP
Supplier:  Enzo Life Sciences BMLLP1040010
Encompass_Preferred
A mitogenic lipid phosphate produced via the action of phospholipase D. Mediates agonist-dependent Raf-1 translocation and activation of the MAP kinase cascade. Activates PKCζ. Alternative name: 1,2-Dioleoyl-sn-glycero-3-phosphate . Na. Purity: ≥98% (GC). Solubility: Soluble in chloroform. Long Term Storage: -20°C.
Catalog No. 50-201-1157