Learn More
YMC America, Inc PA-G guard 120A 5um 10x5mm
SDP
Supplier:  YMC America, Inc PG12S050105WFG
Encompass_Preferred
YMC-Pack PA-G is a normal phase column utilizing amino groups It is useful for separation of acidic oligosaccharides and exhibits higher durability than conventional silica-based amino columns
Catalog No. 50-008-80624