Learn More
YMC America, Inc PA-G guard 120A 5um 20x2.1mm
SDP
Catalog No. 5000880630
Encompass
YMC-Pack PA-G is a normal phase column utilizing amino groups It is useful for separation of acidic oligosaccharides and exhibits higher durability than conventional silica-based amino columns
Catalog No. 50-008-80630 Supplier YMC America, Inc Supplier No. PG12S0502Q1GC
YMC-Pack PA-G is a normal phase column utilizing amino groups It is useful for separation of acidic oligosaccharides and exhibits higher durability than conventional silica-based amino columns