Learn More
YMC America, Inc PA-G guard 120A 5um 20x4mm
SDP
Catalog No. 5000880627
Encompass
YMC-Pack PA-G is a normal phase column utilizing amino groups It is useful for separation of acidic oligosaccharides and exhibits higher durability than conventional silica-based amino columns
Catalog No. 50-008-80627 Supplier YMC America, Inc Supplier No. PG12S050204GC
YMC-Pack PA-G is a normal phase column utilizing amino groups It is useful for separation of acidic oligosaccharides and exhibits higher durability than conventional silica-based amino columns