Learn More
MSC Baldwin Filters Cabin Air Panel Filter, 1 ea
Catalog No. 5000506085
Encompass
Panel Cabin Air Filters help prevent contaminants and pollutants from entering the cabin area. Their space-saving design makes them suitable for a wide variety of heavy-duty trucks and passenger vehicles.
  • Prevents contaminants and pollutants from entering the cabin
  • Space-saving design for diverse vehicle compatibility
  • Includes an attached gasket for a secure seal
  • Dimensions: 9 9/32 in × 8 19/32 in × 25/32 in (235.7 × 218.3 × 19.8 mm)
  • Suitable for applications including Chrysler 200 and Jeep Cherokee
Catalog No. 50-005-06085 Supplier MSC Supplier No. 19374321
Panel Cabin Air Filters help prevent contaminants and pollutants from entering the cabin area. Their space-saving design makes them suitable for a wide variety of heavy-duty trucks and passenger vehicles.
  • Prevents contaminants and pollutants from entering the cabin
  • Space-saving design for diverse vehicle compatibility
  • Includes an attached gasket for a secure seal
  • Dimensions: 9 9/32 in × 8 19/32 in × 25/32 in (235.7 × 218.3 × 19.8 mm)
  • Suitable for applications including Chrysler 200 and Jeep Cherokee