Learn More
MSC Baldwin Cabin Air Element, 239.7 mm Length, 480/Pk.
Catalog No. 5000506437
Encompass
Cabin air elements are designed to prevent contaminants and pollutants from entering the vehicle cabin. Their space-saving design makes them suitable for a wide variety of heavy-duty trucks and passenger vehicles, helping to ensure a safe and comfortable environment.
  • Prevents contaminants and pollutants from entering the cabin area
  • Features a space-saving design
  • Suitable for heavy-duty trucks and passenger vehicles
  • Removes harmful contaminants and pollutants from the air
  • Dimensions: 9 7/16 x 8 x 1 3/16 in (239.7 x 203.2 x 30.2 mm)
  • Applications include Chevrolet Colorado and GMC Canyon
Catalog No. 50-005-06437 Supplier MSC Supplier No. 19374339
Cabin air elements are designed to prevent contaminants and pollutants from entering the vehicle cabin. Their space-saving design makes them suitable for a wide variety of heavy-duty trucks and passenger vehicles, helping to ensure a safe and comfortable environment.
  • Prevents contaminants and pollutants from entering the cabin area
  • Features a space-saving design
  • Suitable for heavy-duty trucks and passenger vehicles
  • Removes harmful contaminants and pollutants from the air
  • Dimensions: 9 7/16 x 8 x 1 3/16 in (239.7 x 203.2 x 30.2 mm)
  • Applications include Chevrolet Colorado and GMC Canyon