Learn More
MSC Baldwin Filters Engine Air Filter, 11 29/32 in, 1 ea
Catalog No. 5000505543
Encompass
This automotive air filter element is designed to protect vehicles and equipment by efficiently removing airborne contaminants from the engine's air intake.
  • Panel design
  • Includes gasket
  • Constructed with cellulose and paper media
  • Rectangular shape
  • Suitable for engine use
  • Vehicle specific fit
  • Compatible with buses and Honda vehicles
  • Manufactured to ISO 9001:2015 standards
Catalog No. 50-005-05543 Supplier MSC Supplier No. 10945434
This automotive air filter element is designed to protect vehicles and equipment by efficiently removing airborne contaminants from the engine's air intake.
  • Panel design
  • Includes gasket
  • Constructed with cellulose and paper media
  • Rectangular shape
  • Suitable for engine use
  • Vehicle specific fit
  • Compatible with buses and Honda vehicles
  • Manufactured to ISO 9001:2015 standards