Learn More
MSC Baldwin Cabin Air Panel Filter, Hyundai Ioniq/Tucson, 1 ea
Catalog No. 5000506155
Encompass
Panel Cabin Air Filters are designed to prevent contaminants and pollutants from entering the vehicle's cabin. This essential component features a space-saving design, making it suitable for a wide range of heavy-duty trucks and passenger vehicles.
  • Helps prevent contaminants and pollutants from entering the cabin area
  • Space-saving design
  • Dimensions: 238.9 x 188.9 x 19.8 mm
  • Engineered for applications in Hyundai Ioniq and Tucson models
Catalog No. 50-005-06155 Supplier MSC Supplier No. 19374610
Panel Cabin Air Filters are designed to prevent contaminants and pollutants from entering the vehicle's cabin. This essential component features a space-saving design, making it suitable for a wide range of heavy-duty trucks and passenger vehicles.
  • Helps prevent contaminants and pollutants from entering the cabin area
  • Space-saving design
  • Dimensions: 238.9 x 188.9 x 19.8 mm
  • Engineered for applications in Hyundai Ioniq and Tucson models