Learn More
MSC Baldwin Axial Seal Air Filter Element, Outer Air Element, For Case Equipment, Attached Gasket
Catalog No. 5000509643
Encompass
Baldwin axial seal air filter elements ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber. These filters feature securely affixed metal end caps with a raised gasket to ensure a proper seal and prevent leaks.
  • Outer air element
  • Designed for Case Equipment
  • Attached gasket
  • Outside diameter 12 1/16 in (306.4 mm)
  • Inside diameter 7 13/16 in (198.4 mm)
  • Length 10 7/16 in (265.1 mm)
Catalog No. 50-005-09643 Supplier MSC Supplier No. 55987127
Baldwin axial seal air filter elements ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber. These filters feature securely affixed metal end caps with a raised gasket to ensure a proper seal and prevent leaks.
  • Outer air element
  • Designed for Case Equipment
  • Attached gasket
  • Outside diameter 12 1/16 in (306.4 mm)
  • Inside diameter 7 13/16 in (198.4 mm)
  • Length 10 7/16 in (265.1 mm)