Learn More
Medchemexpress LLC PA22-2 (IKVAV sequence; laminin A-chain fragment) | 131435-36-4 | 98.96% | 2016.33 | 10 MG
SDP
Catalog No. 5000398011
Encompass_Preferred
PA22-2 (IKVAV sequence; Laminin A-chain fragment) is a biological active peptide. This peptide is derived from mouse laminin a1. Cell matrix substrate constituted with this peptide can promote neurite outgrowth.
  • It is a biological active peptide.
  • It is derived from mouse laminin a1.
  • Cell matrix substrate constituted with this peptide can promote neurite outgrowth.
Catalog No. 50-003-98011 Supplier Medchemexpress LLC Supplier No. HYP535310MG
PA22-2 (IKVAV sequence; Laminin A-chain fragment) is a biological active peptide. This peptide is derived from mouse laminin a1. Cell matrix substrate constituted with this peptide can promote neurite outgrowth.
  • It is a biological active peptide.
  • It is derived from mouse laminin a1.
  • Cell matrix substrate constituted with this peptide can promote neurite outgrowth.