Learn More
MSC Baldwin Axial Seal Air Filter Element, 9 1/2 in OD, 28 7/16 in Length, Inside-Out Flow
Catalog No. 5000509636
Encompass
This air filter element ensures proper equipment function by preventing airborne contaminants from reaching the combustion chamber, contributing to the longevity and performance of the machinery.
  • Designed with inside-out flow direction for effective filtration.
  • Features an outside diameter of 9 1/2 in (241.3 mm).
  • Includes inside diameters of 7/16 in (11.1 mm) and 6 1/2 in (165.1 mm).
  • Measures 28 7/16 in (722.3 mm) in length.
  • Comes with an attached gasket and a furnished gasket (G31-AF).
  • Suitable for use in GMC buses and trucks.
Catalog No. 50-005-09636 Supplier MSC Supplier No. 55990329
This air filter element ensures proper equipment function by preventing airborne contaminants from reaching the combustion chamber, contributing to the longevity and performance of the machinery.
  • Designed with inside-out flow direction for effective filtration.
  • Features an outside diameter of 9 1/2 in (241.3 mm).
  • Includes inside diameters of 7/16 in (11.1 mm) and 6 1/2 in (165.1 mm).
  • Measures 28 7/16 in (722.3 mm) in length.
  • Comes with an attached gasket and a furnished gasket (G31-AF).
  • Suitable for use in GMC buses and trucks.