Learn More
Aviva Systems Biology Corp PA2G4 Antibody - C-terminal region (ARP47601_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47601_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PA2G4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PA2G4. Alternate names: EBP1, HG4-1, p38-2G4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59604