Learn More
Aviva Systems Biology Corp Pa2g4 Antibody - C-terminal region (ARP57879_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57879_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pa2g4. It was validated on Western Blot. . Alternate names: 38kDa, AA672939, Ebp1, PlfapApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-407