Learn More
MSC BALDWIN FILTERS Panel Air Filter, Gasket Attached, 1 ea
Catalog No. 5000505602
Encompass
This panel air filter element is designed for use in engine air filtration systems. It provides efficient filtration and includes a gasket for secure installation.
  • Element only design
  • Panel air element type
  • Gasket attached
  • Overall height: 1 25/32 in
  • Overall length: 13 7/16 in
  • Width: 4 7/16 in to 5 15/16 in
Catalog No. 50-005-05602 Supplier MSC Supplier No. 10943967
This panel air filter element is designed for use in engine air filtration systems. It provides efficient filtration and includes a gasket for secure installation.
  • Element only design
  • Panel air element type
  • Gasket attached
  • Overall height: 1 25/32 in
  • Overall length: 13 7/16 in
  • Width: 4 7/16 in to 5 15/16 in