Learn More
Motic™ PA43 BIO Phase Trinocular Routine Lab Microscope Package
Catalog No. 01257437 Shop All Motic Group Products
Change view
Click to view available options
Camera Type:
6 MP Digital
Catalog No. Camera Type
01-257-437 6 MP Digital
1 options
This item is not returnable. View return policy
Catalog No. 01-257-437 Supplier Motic™ Supplier No. PA43BIOPH1S6
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
The PA43 BIO models have been designed for research use in hospitals and life science laboratories. This package includes the PA43 BIO Trinocular Routine Lab Microscope with 0.5X C-mount camera adapter (PA53 Series), Moticam and Motic™ Analysis Basic 64-bit.
  • Scalable Infinity-Corrected Optical System (CCIS™)
  • Full power transmitted Köhler illumination
  • Phase contrast methods for various microbiology applications
  • Darkfield accessories for blood cell studies for natural therapy
  • Polarizing contrast accessories for protein crystal, tissue, gout and kidney stone observations
  • Environmentally friendly 'ECO' function
  • Consistent, even illumination with IL Function
  • Rackless stage
  • Diopter Adjustment: ±5
Specifications
Magnification Power 10X
Objective Types UC Plan 4X, 100X and UC PH Plan 10X, 40X
Head Style Trinocular
Camera Type 6 MP Digital
Lighting Type Kohler Illumination
Eyepiece WF 10X/22 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov