Learn More
Motic™ PA43 BIO Trinocular Routine Laboratory Microscopes
Catalog No. 01257423 Shop All Motic Group Products
Change view
Click to view available options
Objective Types:
UC Plan 4X and UC PH Plan 10X, 40X, 100X
UC Plan 4X, 100X and UC PH Plan 10X, 40X
UC Plan 4X, 10X, 40X, 100X
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Objective Types
01-257-423 UC Plan 4X, 10X, 40X, 100X
01-257-424 UC Plan 4X, 100X and UC PH Plan 10X, 40X
01-257-425 UC Plan 4X and UC PH Plan 10X, 40X, 100X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-257-423 Supplier Motic™ Supplier No. 1100105500011
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
The PA43 BIOMED combines modularity with ergonomics to improve application imaging workflows. A progression into routine work, this microscope’s robust design and solid footprint does not compromise on optical quality and features.
  • Scalable Infinity-Corrected Optical System (CCIS™)
  • Full power transmitted Köhler illumination
  • Phase contrast methods for various microbiology applications
  • Darkfield accessories for blood cell studies for natural therapy
  • Polarizing contrast accessories for protein crystal, tissue, gout and kidney stone observations
  • Environmentally friendly 'ECO' function
  • Consistent, even illumination with IL Function
  • Rackless stage
  • Diopter Adjustment: ±5
Specifications
Magnification Power 10X
Objective Types UC Plan 4X, 10X, 40X, 100X
Head Style Trinocular
Lighting Type Kohler Illumination
Eyepiece WF 10X/22 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov