Learn More
TARGETMOL CHEMICALS INC PA452 5MG
SDP
Catalog No. 502581177
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PA452 is a selective antagonist of retinoic X receptor (RXR) and suppresses the effect of Retinoic acid on Th1/Th2 development. purity: 96%
Catalog No. 50-258-1177 Supplier TARGETMOL CHEMICALS INC Supplier No. T164255MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PA452 is a selective antagonist of retinoic X receptor (RXR) and suppresses the effect of Retinoic acid on Th1/Th2 development. purity: 96%