Learn More
Motic™ PA53 MET-3D Materials Motorized Z-Drive Digital Microscope with Analysis Professional Software Suite
Catalog No. 01257431 Shop All Motic Group Products
Change view
Click to view available options
Head Style:
Trinocular
Catalog No. Head Style
01-257-431 Trinocular
1 options
This item is not returnable. View return policy
Catalog No. 01-257-431 Supplier Motic™ Supplier No. 1100104600871
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
The PA53MET-3D Profiler is a modular high-end materials microscope with a fully motorized Z-axis, XY stage and objective nosepiece. Motic™’s Analysis Software adds highly accurate measurements in X, Y, and Z planes at submicron levels.
  • Scalable Infinity-Corrected Optical System (CCIS™)
  • Superior optical performance with newly designed Semi-Apochromatic objectives
  • Interference Contrast (DIC) concept incident illumination
  • Established contrast methods
  • Multiple degrees of motorization in X, Y, and Z
  • Harmonized Brightfield/Darkfield switch
  • Transmitted Light: LB daylight filter (Halogen lamp)
  • Diopter adjustable
Specifications
Magnification Power 10X
Objective Types S-APO Plan 5X, 10X, 20X, 50X
Head Style Trinocular
Lighting Type Halogen
Eyepiece WF 10X/25 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov