Learn More
Motic™ PA53 MET Trinocular Upright Materials Microscope
Catalog No. 01257428 Shop All Motic Group Products
Change view
Click to view available options
Lighting Type:
Halogen
Reflected Illumination
Reflected and Transmitted Illumination
Model:
PA53 MET
PA53 MET BD
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lighting Type Model
01-257-428 Reflected Illumination PA53 MET BD
01-257-429 Reflected and Transmitted Illumination PA53 MET
01-257-430 Reflected and Transmitted Illumination PA53 MET BD
01-257-427 Halogen PA53 MET
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 01-257-428 Supplier Motic™ Supplier No. 1100104600881
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
A combination of technology and creativity, the PA53 MET series offers superior optical performance as well as an advanced ergonomic design, futuristic digital capabilities, and innovative modules ideal for metallurgy.
  • Scalable Infinity-Corrected Optical System (CCIS™)
  • Superior optical performance with newly designed Semi-Apochromatic objectives
  • Interference Contrast (DIC) concept incident illumination
  • Established contrast methods
  • Multiple degrees of motorization in X, Y, and Z
  • Harmonized Brightfield/Darkfield switch
  • Transmitted Light: LB daylight filter (Halogen lamp)
  • Diopter adjustable
Specifications
Magnification Power 10X
Objective Types S-APO BD Plan 5X, 10X, 20X, 50X
Model PA53 MET BD
Head Style Trinocular
Lighting Type Reflected Illumination
Eyepiece WF 10X/25 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov