Learn More
MSC Baldwin Filters Air Filter Element, 11 17/32 in Height, 12 13/16 in Outside Diameter
Catalog No. 5000508196
Encompass
This air filter element is engineered for engine applications, providing efficient filtration. It features a compact design with specific dimensions to ensure optimal performance in compatible systems.
  • Element only air filter design.
  • Height of 11 17/32 in.
  • Outside diameter of 12 13/16 in.
  • Includes two attached gaskets.
  • Suitable for engine air filtration.
Catalog No. 50-005-08196 Supplier MSC Supplier No. 55873970
This air filter element is engineered for engine applications, providing efficient filtration. It features a compact design with specific dimensions to ensure optimal performance in compatible systems.
  • Element only air filter design.
  • Height of 11 17/32 in.
  • Outside diameter of 12 13/16 in.
  • Includes two attached gaskets.
  • Suitable for engine air filtration.