Learn More
Honeywell™ North™ Primair™ 700 Series Powered Air Purifying Respirator
PAPR belt-mounted platform is ergonomically designed to distribute weight evenly to prevent slipping.
Supplier:  Honeywell™ PA701HED
 View more versions of this product
Catalog No. 19-085-498
This item is not returnable. View return policy
Description
Description
  • Assembly includes connected blower, battery, battery charger, belt
  • PA701 HE also includes comfort pad, bonus P100 filter and optional filter cover
  • Belt option includes nylon or decontamation
  • Side mounted battery locks into place to reduce accidental disconnections.
  • Ergonomic belt distributes weight evenly and prevents slipping
  • Adjustable nylon straps on belt
  • Battery Run Time: 8 Hours
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8 hours
Decontamination
PAPR Assembly, No Headtop
Primair™
1 Ea.
Lithium-ion
Twist and lock connector
Blower, battery, battery charger, and decon belt
Belt-mounted
6 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.