Learn More
OriGene pAAV-AC-RFP AAV Gene Expression Vector, 10 ug
SDP
Catalog No. NC2938438
Encompass_Preferred
OriGene’s TrueORF clones can be shuttled into this vector via precisionShuttle, a simple cut-and-ligation process.The ORF cloned in this vector can be expressed containing c-terminal tRFP.BGI_F Primer can be used to sequence the target sequence after cloning.RFP_R Primer can be used to sequence the target sequence after cloning.
Catalog No. NC2938438 Supplier OriGene Supplier No. PS100091
OriGene’s TrueORF clones can be shuttled into this vector via precisionShuttle, a simple cut-and-ligation process.The ORF cloned in this vector can be expressed containing c-terminal tRFP.BGI_F Primer can be used to sequence the target sequence after cloning.RFP_R Primer can be used to sequence the target sequence after cloning.