Learn More
Cell Biolabs Inc pAAV-DJ Vector, 10 µg
Catalog No. 50673893
Encompass_Preferred
AAV-DJ is a hybrid AAV serotype developed from the recombination of 8 different native AAV serotypes. AAV-DJ viruses can infect a broad range of target cell types with high transduction efficieny. Cell Biolabs' pAAV-DJ vector serves as the Rep-Cap vector to be co-transfected with other AAV vectors into 293 cells for viral packaging. FOR SALE TO ACADEMIC, GOVERNMENT AND NON-PROFIT INSTITUTIONS ONLY.
Catalog No. 50-673-893 Supplier Cell Biolabs Inc Supplier No. VPK420DJ
AAV-DJ is a hybrid AAV serotype developed from the recombination of 8 different native AAV serotypes. AAV-DJ viruses can infect a broad range of target cell types with high transduction efficieny. Cell Biolabs' pAAV-DJ vector serves as the Rep-Cap vector to be co-transfected with other AAV vectors into 293 cells for viral packaging. FOR SALE TO ACADEMIC, GOVERNMENT AND NON-PROFIT INSTITUTIONS ONLY.