Learn More
Cell Biolabs Inc pAAV-GFP Control Plasmid, 10 µg
Catalog No. 50672457
Encompass_Preferred
Cell Biolabs' pAAV-GFP Control Vector may be co-transfected with AAV packaging plasmids, such as an AAV Helper Free Packaging System, to generated recombinant adeno-associated virus. The resulting virus may be used as a transduction control.
Catalog No. 50-672-457 Supplier Cell Biolabs Inc Supplier No. AAV400
Cell Biolabs' pAAV-GFP Control Vector may be co-transfected with AAV packaging plasmids, such as an AAV Helper Free Packaging System, to generated recombinant adeno-associated virus. The resulting virus may be used as a transduction control.