Learn More
Cell Biolabs Inc pAAV-RC2 Vector, 10 µg
Catalog No. 50673896
Encompass_Preferred
Cell Biolabs' pAAV-RC2 Vector contains the rep and cap genes required to generated recombinant AAV of serotype 1. Co-transfect this vector with other packaging plasmids and an expression vector into 293 cells for AAV-2 packaging.
Catalog No. 50-673-896 Supplier Cell Biolabs Inc Supplier No. VPK422
 Restricted Item
Add to Cart
Add to Cart
Cell Biolabs' pAAV-RC2 Vector contains the rep and cap genes required to generated recombinant AAV of serotype 1. Co-transfect this vector with other packaging plasmids and an expression vector into 293 cells for AAV-2 packaging.