Learn More
Cell Biolabs Inc pAAV-RC6 Vector, 10 µg
Supplier:  Cell Biolabs Inc VPK426
Encompass_Preferred
Cell Biolabs' pAAV-RC6 Vector contains the rep and cap genes required to generated recombinant AAV of serotype 6. Co-transfect this vector with other packaging plasmids and an expression vector into 293 cells for AAV-6 packaging.
Catalog No. 50-673-900