Learn More
Adam Equipment PAB 124e - Polaris Analytical Balances - Capacity: 120g, Readability: 0.0001g
Supplier:  Adam Equipment PAB 124E
Encompass_Preferred
Polaris semi-micro and analytical balances deliver exceptional precision and reliability for demanding lab and industrial applications. A brightly illuminated weighing chamber and intuitive multi-line display provide fast, accurate results with clear guidance. The color-changing backlight streamlines checkweighing by instantly indicating under, over, or within limits—reducing errors and improving efficiency.Designed for regulated environments, Polaris supports GLP and GMP compliance for dependable data tracking. A large grade 304 stainless steel pan ensures easy cleaning, while durable ABS construction stands up to industrial use. An integrated airflow channel minimizes environmental interference for enhanced accuracy.With built-in Bluetooth, RJ45 Ethernet, RS-232, and USB connectivity, Polaris enables seamless data integration. A Kensington lock slot adds security, making it a powerful, user-friendly solution for precision weighing.
Catalog No. NC3964980