Learn More
Adam Equipment PAB 125i - Polaris Semi-Micro Analytical Balances - Capacity: 80g / 120g, Readability: 0.00001g / 0.0001g
Catalog No. NC3964968
Encompass_Preferred
Polaris semi-micro and analytical balances deliver precision and reliability for laboratory and industrial environments. A brightly illuminated weighing chamber and multi-line display provide clear guidance and fast, accurate results. The color-changing backlight simplifies checkweighing by instantly indicating under, over, or acceptable limits, improving efficiency and reducing errors.Designed for regulated settings, Polaris supports GLP and GMP compliance for reliable data tracking. A large grade 304 stainless steel pan ensures easy cleaning, while durable ABS housing withstands industrial use. A unique airflow diversion channel minimizes environmental interference for improved accuracy.With built-in Bluetooth, RJ45 Ethernet, RS-232, and USB, Polaris enables seamless data transfer and integration. A Kensington-type lock slot adds security, making it a dependable, user-friendly solution for precise weighing applications.
Catalog No. NC3964968 Supplier Adam Equipment Supplier No. PAB 125I
Polaris semi-micro and analytical balances deliver precision and reliability for laboratory and industrial environments. A brightly illuminated weighing chamber and multi-line display provide clear guidance and fast, accurate results. The color-changing backlight simplifies checkweighing by instantly indicating under, over, or acceptable limits, improving efficiency and reducing errors.Designed for regulated settings, Polaris supports GLP and GMP compliance for reliable data tracking. A large grade 304 stainless steel pan ensures easy cleaning, while durable ABS housing withstands industrial use. A unique airflow diversion channel minimizes environmental interference for improved accuracy.With built-in Bluetooth, RJ45 Ethernet, RS-232, and USB, Polaris enables seamless data transfer and integration. A Kensington-type lock slot adds security, making it a dependable, user-friendly solution for precise weighing applications.