Learn More
Genscript Corporation Avi-tag Antibody, pAb, Rabbit
Supplier:  Genscript Corporation A0067440
Encompass_Preferred
Well-characterized antibodies for epitope tags have been widely used in the study of protein expression in various systems.The Avi tag is a biotin-acceptor peptide1, GLNDIFEAQKIEWHE.The 15-residue peptide served as a substrate mimic for biotin ligase (BirA), which usually recognizes the much larger protein domain.GenScript Rabbit Anti-Avi-tag Polyclonal Antibody is developed in rabbit using a synthetic Avi tag peptide conjugated to KLH.
Catalog No. 50-272-909