Learn More
Sino Biological PABPC1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 105506T44100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PABPC1, and purified by antigen affinity chromatography.
Catalog No. 50-224-6329